missing translation for 'onlineSavingsMsg'
Få mere at vide

di-Phosphorus Pentoxide, Supelco™

Product Code. 15823910
Change view
Click to view available options
Mængde:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
15823910 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15823910 Supplier Supelco Supplier No. 1005401000
 Begrænset vare
Forventet forsendelse: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

Extra pure

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Di-Phosphorus Pentoxide
Smeltepunkt 420°C (Closed Capillary Tube)
CAS 1314-56-3
Mængde 1 kg
MDL nummer MFCD00148880
Synonym di-Phosphorus pentoxide, Phosphoric anhydride, di-Phosphorus pentoxide
Molekylvægt (g/mol) 141.94
Grad Research
Tæthed 2.28 g/cm3 at 20 to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.