Få mere at vide
Di-n-undecyl Phthalate-3,4,5,6-d4, CDN
Artikelnummer. 30320755
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320755 0.05 g -
30319057 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320755 Leverandør CDN Leverandørnr. CDND76670.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Diundecyl Phthalate-3,4,5,6-D4
CAS 2708278-18-4
Molekylær formel C30 D4 H46 O4
Mængde 0.05 g
Synonym Di-n-undecyl Phthalate-3,4,5,6-d4, Diundecyl phthalate-3,4,5,6-d4, Phthalic Acid-d4 Bis-undecyl Ester, Diundecyl Phthalate-3,4,5,6-D4
InChI formel InChI=1 S/C30H50O4/c1-3-5-7-9-11-13-15-17-21-25-33-29(31)27-23-19-20-24-28(27)30(32)34-26-22-18-16-14-12-10-8-6-4-2/h19-20,23-24 H,3-18,21-22,25-26H2,1-2H3/i19D,20 D,23 D,24 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCCCCCCC)c(C(=O)OCCCCCCCCCCC)c1[2 H]
IUPAC navn diundecyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 478.7403
Formel vægt 478.396 g/mol
Procent renhed 99 atom % D, min 96% Chemical Purity