Få mere at vide
Di-n-pentyl Phthalate-3,4,5,6-d4, CDN
Artikelnummer. 30313121
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30313121 0.25 g
30320488 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313121 Leverandør CDN Leverandørnr. CDND55050.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic acid, bis-n-pentyl ester D4
CAS 358730-89-9
Molekylær formel C18 2H4 H22 O4
Mængde 0.25 g
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dipentyl ester (9 CI), Phthalic acid, bis-n-pentyl ester D4, Amyl Phthalate-d4, Di-n-pentyl Phthalate-d4, Phthalic Acid Di-n-pentyl Ester (3,4,5,6)-D4
InChI formel InChI=1 S/C18H26O4/c1-3-5-9-13-21-17(19)15-11-7-8-12-16(15)18(20)22-14-10-6-4-2/h7-8,11-12 H,3-6,9-10,13-14H2,1-2H3/i7D,8 D,11 D,12 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCC)c(C(=O)OCCCCC)c1[2 H]
IUPAC navn dipentyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 310.42
Formel vægt 310.2082 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity