Få mere at vide
Di-n-octyltin bis(2-ethylhexyl thioglycolate), Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Plastic additive 23 |
| CAS | 15571-58-1 |
| Analyt- eller komponentnavne | Plastic additive 23 |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C36 H72 O4 S2 Sn |
| Certificeringer/compliance | ISO/IEC 17025 |
| Synonym | 10-Ethyl-4,4-dioctyl-7-oxo-8-oxa-3,5-dithia-4-stannatetradecanoic acid 2-ethylhexyl ester, [(Dioctylstannylene)dithio]diacetic acid bis(2-ethylhexyl) ester, Bis[(carboxymethyl)thio]dioctyltin bis(2-ethylhexyl) ester, 17MOK, Advastab 17MOK, Advastab 17MOL, Bis(2-ethylhexyl thioglycolato)dioctyltin, Bis(2-ethylhexyl) [(dioctylstannylene)dithio]diacetate, CAT 214, Dioctyltin bis(2-ethylhexyl thioglycolate), KS 2000A10, Reatinor 847, Thermolite 895, Plastic additive 23 (EP) |
| InChI formel | InChI=1S/2C10H20O2S.2C8H17.Sn/c2*1-3-5-6-9(4-2)7-12-10(11)8-13;2*1-3-5-7-8-6-4-2;/h2*9,13H,3-8H2,1-2H3;2*1,3-8H2,2H3;/q;;;;+2/p-2 |
| Løsningstype | Neat |
| SMIL | CCCCCCCC[Sn](CCCCCCCC)(SCC(=O)OCC(CC)CCCC)SCC(=O)OCC(CC)CCCC |
| Vis mere |