missing translation for 'onlineSavingsMsg'
Learn More

Di-n-octyl sulfide, 97%, Thermo Scientific Chemicals

Catalog Number p-7023925
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
2690-08-6
MFCD00009566
This item is not returnable. View return policy
2690-08-6
MFCD00009566

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Solubility in water, 0.004678 mg/L @ 25°C (est)

Notes
Air & Moisture Sensitive. Store away from acids, oxidizing agents, air, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 0°C to 1°C
Density 0.844
Boiling Point 179°C to 180°C (10 mmHg)
Flash Point 160°C (320°F)
Odor Stench
Molecular Formula C16H34S
Refractive Index 1.4624
Quantity 25 g
Beilstein 1701624
Synonym dioctyl sulfide, di-n-octyl sulfide, octane, 1,1'-thiobis, octyl sulfide, n-octyl sulfide, dioctyl thioether, 1-octylsulfanyl octane, 9-thiaheptadecane, di-n-octylsulfide, dioctylsulfane
InChI Key LOXRGHGHQYWXJK-UHFFFAOYSA-N
SMILES CCCCCCCCSCCCCCCCC
IUPAC Name 1-octylsulfanyloctane
Molecular Weight (g/mol) 258.51
PubChem CID 75901
Formula Weight 258.51
Percent Purity 97%
Chemical Name or Material Di-n-octyl sulfide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'