Få mere at vide
Di-n-octyl Phthalate-3,4,5,6-d4, CDN
Artikelnummer. 30321874
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321874 0.1 g 0.1g
30317954 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321874 Leverandør CDN Leverandørnr. CDND22910.1G
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic acid, bis-n-octyl ester-3,4,5,6-D4
CAS 93952-13-7
Molekylær formel C24 2H4 H34 O4
Mængde 0.1 g
Synonym Di-n-octyl Phthalate-d4, Phthalic acid bis-n-octyl ester-3,4,5,6-D4, 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dioctyl ester (9 CI), Phthalic Acid Di-n-octyl Ester (3,4,5,6)-D4
InChI formel InChI=1 S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18 H,3-12,15-16,19-20H2,1-2H3/i13D,14 D,17 D,18 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCCCC)c(C(=O)OCCCCCCCC)c1[2 H]
IUPAC navn dioctyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 394.58
Formel vægt 394.3021 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity