missing translation for 'onlineSavingsMsg'
Få mere at vide

Di-n-dodecyl-d25-dimethylammonium Bromide (mono-n-dodecyl-d25), TRC

Artikelnummer. 30377898
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30377898 2.5 mg 2.5mg
30380279 5 mg -
30386249 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30377898 Supplier Toronto Research Chemicals Supplier No. TRCD4417462.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 918897-30-0
Molekylær formel C26 D25 H31 N . Br
Molekylvægt (g/mol) 487.7876
Synonym 1-Dodecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25-aminium, N-dodecyl-N,N-dimethyl-, bromide (1:1) (ACI), N-Dodecyl-N,N-dimethyl-1-dodecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25-aminium bromide, Di-n-dodecyl-d25-dimethylammonium Bromide (mono-n-dodecyl-d25), Didodecyldimethylammonium-D25 (mono-dodecyl-D25) bromide, N-Dodecyl-N-(dodecyl-d25)-N,N-dimethylammonium bromide
IUPAC navn dodecyl-dimethyl-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-pentacosadeuteriododecyl)azanium;bromide
SMIL [Br-].[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[N+](C)(C)CCCCCCCCCCCC

Specifications

Kemisk navn eller materiale Di-n-dodecyl-d25-dimethylammonium Bromide (mono-n-dodecyl-d25)
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C26H56N.BrH/c1-5-7-9-11-13-15-17-19-21-23-25-27(3,4)26-24-22-20-18-16-14-12-10-8-6-2;/h5-26H2,1-4H3;1H/q+1;/p-1/i1D3,5D2,7D2,9D2,11D2,13D2,15D2,17D2,19D2,21D2,23D2,25D2;
Formel vægt 486.5165
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.