Få mere at vide
Di-n-butyl Phthalate-d22, TRC
Artikelnummer. 30375525
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
30375525 2 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375525 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4294962MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Di-n-butyl Phthalate-d22
CAS 358731-15-4
Molekylær formel C16 D22 O4
Mængde 2 mg
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, di(butyl-d9) ester (9CI), Di-(n-butyl-d9) Phthalate-3,4,5,6-d4, Phthalic Acid Bis-butyl Ester D22, Dibutyl Phthalate-d22, Dibutyl Phthalate-D22, 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid di(butyl-d9) ester
InChI formel InChI=1S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D,8D,9D,10D,11D2,12D2
SMIL [2H]c1c([2H])c([2H])c(C(=O)OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])c(C(=O)OC([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])c1[2H]
IUPAC navn bis(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 300.479
Formel vægt 300.2899