Få mere at vide
Di-n-butyl Phthalate-3,4,5,6-d4, CDN
Artikelnummer. 30318335
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30318335 0.25 g -
30319249 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318335 Leverandør CDN Leverandørnr. CDND23100.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic acid, bis-butyl ester D4
CAS 93952-11-5
Molekylær formel C16 2H4 H18 O4
Mængde 0.25 g
InChI formel InChI=1 S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10 H,3-6,11-12H2,1-2H3/i7D,8 D,9 D,10 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCC)c(C(=O)OCCCC)c1[2 H]
IUPAC navn dibutyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 282.37
Formel vægt 282.1769 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity