Få mere at vide
Di-m-tolyl Phosphate, TRC
Artikelnummer. 30378432
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30378432 100 mg
1 options
Artikelnummer. 30378432 Leverandør Toronto Research Chemicals Leverandørnr. TRCD436800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Di-m-tolyl Phosphate
CAS 36400-46-1
Molekylær formel C14H15O4P
Mængde 100 mg
Synonym Phosphoric Acid Bis(3-methylphenyl) Ester, m-Tolyl Phosphate ((C7H7O)2(HO)PO), Di-m-cresyl Phosphate
InChI formel InChI=1S/C14H15O4P/c1-11-5-3-7-13(9-11)17-19(15,16)18-14-8-4-6-12(2)10-14/h3-10H,1-2H3,(H,15,16)
SMIL Cc1cccc(OP(=O)(O)Oc2cccc(C)c2)c1
IUPAC navn bis(3-methylphenyl) hydrogen phosphate
Molekylvægt (g/mol) 278.24
Formel vægt 278.0708