Få mere at vide
Di-iso-propyl-d14-amine HCl, CDN
Artikelnummer. 30316444
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316444 0.1 g 0.1g
30310925 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316444 Leverandør CDN Leverandørnr. CDND58020.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N,N-Diisopropylamine-d14, Hydrochloride
CAS 1219803-88-9
Molekylær formel C6 2H14 H N . Cl H
Mængde 0.1 g
InChI formel InChI=1 S/C6H15N.ClH/c1-5(2)7-6(3)4;/h5-7 H,1-4H3;1 H/i1D3,2D3,3D3,4D3,5 D,6 D;
SMIL Cl.[2 H]C([2 H])([2 H])C([2 H])(NC([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,2,3,3,3-heptadeuterio-N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)propan-2-amine;hydrochloride
Molekylvægt (g/mol) 151.74
Formel vægt 151.185 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity