Få mere at vide
Di-iso-butyl Phthalate-3,4,5,6-d4, CDN
Artikelnummer. 30321901
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321901 0.1 g
30325314 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321901 Leverandør CDN Leverandørnr. CDND55040.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Phthalic acid, bis-iso-butyl ester D4
CAS 358730-88-8
Molekylær formel C16 2H4 H18 O4
Mængde 0.1 g
Synonym 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, bis(2-methylpropyl) ester (9 CI), Phthalic acid, bis-iso-butyl ester D4, Diisobutyl Phthalate-d4, Phthalic Acid Diisobutyl Ester (3,4,5,6)-D4
InChI formel InChI=1 S/C16H22O4/c1-11(2)9-19-15(17)13-7-5-6-8-14(13)16(18)20-10-12(3)4/h5-8,11-12 H,9-10H2,1-4H3/i5D,6 D,7 D,8 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)OCC(C)C)c(C(=O)OCC(C)C)c1[2 H]
IUPAC navn bis(2-methylpropyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate
Molekylvægt (g/mol) 282.37
Formel vægt 282.1769 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity