missing translation for 'onlineSavingsMsg'
Learn More

Di-μ-hydroxo-bis-[(N,N,N',N'-tetramethylethylenediamine)copper(II)]chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-120599
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
30698-64-7
C12H36Cl2Cu2N4O2
466.44
MFCD01321163
VVXKYYDFGPZSOZ-UHFFFAOYSA-L
cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride
11190467
chlorocopper;N,N,N',N'-tetramethylethane-1,2-diamine;dihydrate
O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C
This item is not returnable. View return policy
30698-64-7
C12H36Cl2Cu2N4O2
466.44
MFCD01321163
VVXKYYDFGPZSOZ-UHFFFAOYSA-L
cu-tmeda catalyst, di-mu-hydroxo-bis n,n,n'.n'-tetramethylethylenediamine copper ii chloride, cu-tmedacatalyst, bis chlorocopperol ; bis temed, di-hydroxo-bis n,n,n',n'-tetramethylethylenediamine copper ii chloride, chlorohydroxo n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine copper ii, copper ii chloride hydroxide n,n,n inverted exclamation marka,n inverted exclamation marka-tetrametylethylenediamine complex dimer, di-, i-hydroxo-bis n,n,n inverted exclamation marka,n inverted exclamation marka-tetramethylethylenediamine copper ii chloride, i-hydroxo-bis n,n,n inverted exclamation marka.n inverted exclamation marka-tetramethylethylenediamine copper ii chloride
11190467
chlorocopper;N,N,N',N'-tetramethylethane-1,2-diamine;dihydrate
O.O.Cl[Cu].Cl[Cu].CN(C)CCN(C)C.CN(C)CCN(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 136.0°C to 138.0°C
Color Purple
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 464.42
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Di-μ-hydroxo-bis-[(N,N,N′,N′-tetramethylethylenediamine)copper(II)]chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'