Få mere at vide
Di-Beta-carbethoxyethyl-d8-methylamine, TRC
Artikelnummer. 30370123
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30370123 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30370123 Leverandør Toronto Research Chemicals Leverandørnr. TRCD430652500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Di-beta-carbethoxyethyl-d8-methylamine
CAS 1189723-57-6
Molekylær formel C11 2H8 H13 N O4
Mængde 500 mg
InChI formel InChI=1S/C11H21NO4/c1-4-15-10(13)6-8-12(3)9-7-11(14)16-5-2/h4-9H2,1-3H3/i6D2,7D2,8D2,9D2
SMIL [2H]C([2H])(N(C)C([2H])([2H])C([2H])([2H])C(=O)OCC)C([2H])([2H])C(=O)OCC
IUPAC navn ethyl 2,2,3,3-tetradeuterio-3-[methyl-(1,1,2,2-tetradeuterio-3-ethoxy-3-oxopropyl)amino]propanoate
Molekylvægt (g/mol) 239.34
Formel vægt 239.1973