Få mere at vide
Di-Beta,Beta'-Chloroethylphosphoric Acid-d8, TRC
Artikelnummer. 30365341
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30365341 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30365341 Leverandør Toronto Research Chemicals Leverandørnr. TRCC36679210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Di-beta,beta'-Chloroethylphosphoric Acid-d8
CAS 1477495-02-5
Renhedsgrad noter HPLC
Molekylær formel C4HD8Cl2O4P
Mængde 10 mg
Synonym Bis(2-chloroethyl) Hydrogen Phosphate-d8, Bis(2-chloroethyl) Phosphate-d8, Bis(chloroethyl) Phosphate-d8, Bis(β-chloroethyl) Phosphate-d8, Bis(β-chloroethyl) Phosphoric Acid-d8, Di-2-Chloroethyl Phosphate-d8
InChI formel InChI=1S/C4H9Cl2O4P/c5-1-3-9-11(7,8)10-4-2-6/h1-4H2,(H,7,8)/i1D2,2D2,3D2,4D2
SMIL O=P(OC([2H])([2H])C([2H])([2H])Cl)(OC([2H])([2H])C([2H])([2H])Cl)O
IUPAC navn bis(2-chloro-1,1,2,2-tetradeuterioethyl) hydrogen phosphate
Molekylvægt (g/mol) 231.04
Formel vægt 230.0118
Procent renhed >95