missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Ammonium Hydrogen Orthophosphate, Certified AR for analysis, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Description
Di-Ammonium Hydrogen Orthophosphate, Certified AR for analysis
Specifications
Color | White |
pH | 7.9 to 8.3 |
Physical Form | Crystalline Solid |
Quantity | 500 g |
Formula Weight | 132.06g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'