Få mere at vide
Di(4-nitrophenyl)phosphoryl Chloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Di(4-nitrophenyl)phosphoryl Chloride |
| CAS | 6546-97-0 |
| Molekylær formel | C12 H8 Cl N2 O7 P |
| Mængde | 1 g |
| Synonym | Phosphorochloridic acid, bis(4-nitrophenyl) ester, Phenol, p-nitro-, phosphorochloridate (7CI), Phosphorochloridic acid, bis(p-nitrophenyl) ester (6CI,8CI), Bis(p-nitrophenyl) phosphorochloridate, Di(4-nitrophenyl)phosphoryl chloride, Phosphorochloridic acid bis(4-nitrophenyl) ester |
| InChI formel | InChI=1S/C12H8ClN2O7P/c13-23(20,21-11-5-1-9(2-6-11)14(16)17)22-12-7-3-10(4-8-12)15(18)19/h1-8H |
| SMIL | [O-][N+](=O)c1ccc(OP(=O)(Cl)Oc2ccc(cc2)[N+](=O)[O-])cc1 |
| IUPAC navn | 1-[chloro-(4-nitrophenoxy)phosphoryl]oxy-4-nitrobenzene |
| Molekylvægt (g/mol) | 358.63 |
| Formel vægt | 357.9758 |