missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DHRS7 Recombinant Protein Antigen
Recombinant Protein
Brand: Novus Biologicals™ NBP3-17357PEP
Additional Details : Weight : 0.00970kg
Specifications
Antibody Competition | |
51635 | |
100 μg | |
dehydrogenase/reductase (SDR family) member 7,2310016E22Rik, dehydrogenase/reductase SDR family member 7, DHRS7A, EC 1.1, retDSR4, Retinal short-chain dehydrogenase/reductase 4, RETSDR4, SDR34C1, short chain dehydrogenase/reductase family 34C, member 1 | |
Lipid and Metabolism | |
DHRS7 |
PBS and 1M Urea, pH 7.4. | |
M.W. Theoretical: 25 kDa | |
Store at -20°C. Avoid freeze-thaw cycles. | |
DHRS7 | |
>80% by SDS-PAGE and Coomassie blue staining |