Få mere at vide
(DHQD)2PHAL, TRC
Artikelnummer. 30359669
Skift visning
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30359669 5 g
30352449 500 mg
30356903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30359669 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2999055G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 140853-10-7
Molekylær formel C48H54N6O4
Molekylvægt (g/mol) 778.98
Synonym Hydroquinidine 1,4-Phthalazinediyl Ether, SYKINH-61, 1,4-Bis(dihydroquinidine)phthalazine, 1,4-Bis(9'-O-dihydroquinidyl)phthalazine, 1,4-Bis(9-O-dihydroquinidyl)phthalazine, 1,4-Bis(9-O-dihydroquinidinyl)phthalazine, (9S)-(9''S)-9,9''-[1,4-Phthalazinediylbis(oxy)]bis[10,11-dihydro-6'-methoxycinchonan]
SMIL CC[C@H]1CN2CC[C@H]1C[C@@H]2[C@@H](Oc3nnc(O[C@H]([C@H]4C[C@@H]5CCN4C[C@@H]5CC)c6ccnc7ccc(OC)cc67)c8ccccc38)c9ccnc%10ccc(OC)cc9%10
Tekniske data
Kemisk navn eller materiale (DHQD)2PHAL
Mængde 5 g
InChI formel InChI=1S/C48H54N6O4/c1-5-29-27-53-21-17-31(29)23-43(53)45(35-15-19-49-41-13-11-33(55-3)25-39(35)41)57-47-37-9-7-8-10-38(37)48(52-51-47)58-46(44-24-32-18-22-54(44)28-30(32)6-2)36-16-20-50-42-14-12-34(56-4)26-40(36)42/h7-16,19-20,25-26,29-32,43-46H,5-6,17-18,21-24,27-28H2,1-4H3/t29-,30-,31-,32-,43+,44+,45-,46-/m0/s1
Formel vægt 778.4207