Få mere at vide
(DHQ)2PHAL, TRC
Artikelnummer. 30180049
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180049 500 mg
30180113 100 mg
30179991 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180049 Leverandør Toronto Research Chemicals Leverandørnr. TRCD299920500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 140924-50-1
Molekylær formel C48H54N6O4
Molekylvægt (g/mol) 778.98
Synonym 1,4-Bis(9-O-dihydroquininyl)phthalazine, 1,4-Bis(9-O-dihydroquinyl)phthalazine, 1,4-Bis(dihydroquinine)phthalazine, (8alpha,9R)-(8' 'alpha,9' 'R)-9,9' '-[1,4-Phthalazinediylbis(oxy)]bis[10,11-dihydro-6'-methoxycinchonan];
SMIL CC[C@H]1CN2CC[C@H]1C[C@H]2[C@H](Oc3nnc(O[C@@H]([C@@H]4C[C@@H]5CCN4C[C@@H]5CC)c6ccnc7ccc(OC)cc67)c8ccccc38)c9ccnc%10ccc(OC)cc9%10
Tekniske data
Kemisk navn eller materiale (DHQ)2PHAL
Mængde 500 mg
InChI formel InChI=1S/C48H54N6O4/c1-5-29-27-53-21-17-31(29)23-43(53)45(35-15-19-49-41-13-11-33(55-3)25-39(35)41)57-47-37-9-7-8-10-38(37)48(52-51-47)58-46(44-24-32-18-22-54(44)28-30(32)6-2)36-16-20-50-42-14-12-34(56-4)26-40(36)42/h7-16,19-20,25-26,29-32,43-46H,5-6,17-18,21-24,27-28H2,1-4H3/t29-,30+,31-,32+,43-,44-,45-,46-/m1/s1
Formel vægt 778.4207