Få mere at vide
DHPMA, TRC
Artikelnummer. 30136309
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136309 250 mg
30134323 500 mg
30004179 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136309 Leverandør Toronto Research Chemicals Leverandørnr. TRCD965260250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 23255-15-4
Molekylær formel C8H15NO5S
Molekylvægt (g/mol) 237.27
Synonym N-Acetyl-3-[(2,3-dihydroxypropyl)thio]-alanine
IUPAC navn N-acetyl-S-(2,3-dihydroxypropyl)cysteine
SMIL CC(=O)NC(CSCC(O)CO)C(=O)O
Tekniske data
Kemisk navn eller materiale DHPMA
InChI formel InChI=1S/C8H15NO5S/c1-5(11)9-7(8(13)14)4-15-3-6(12)2-10/h6-7,10,12H,2-4H2,1H3,(H,9,11)(H,13,14)
Mængde 250 mg
Formel vægt 237.0671