Få mere at vide
Dextrosimendan 6-Phenyl, TRC
Artikelnummer. 30470325
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30470325 250 mg
30474398 100 mg
30476667 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30470325 Leverandør Toronto Research Chemicals Leverandørnr. TRCL589170250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2290570-69-1
Molekylær formel C11H12N2O
Molekylvægt (g/mol) 188.23
Synonym (5S)-4,5-Dihydro-5-methyl-6-phenyl-3(2H)-pyridazinone
IUPAC navn (4S)-4-methyl-3-phenyl-4,5-dihydro-1H-pyridazin-6-one
SMIL C[C@H]1CC(=O)NN=C1c2ccccc2
Tekniske data
Kemisk navn eller materiale Dextrosimendan 6-Phenyl
Mængde 250 mg
InChI formel InChI=1S/C11H12N2O/c1-8-7-10(14)12-13-11(8)9-5-3-2-4-6-9/h2-6,8H,7H2,1H3,(H,12,14)/t8-/m0/s1
Formel vægt 188.095