missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextrorphan tartrate, Dr. Ehrenstorfer
Specifications
Specifications
| Kemisk navn eller materiale | Dextrorphan tartrate |
| CAS | 143-98-6 |
| Analyt- eller komponentnavne | Dextrorphan tartrate |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C17 H23 N O . C4 H6 O6 |
| Certificeringer/compliance | ISO/IEC 17025 |
| Synonym | ent-17-Methylmorphinan-3-ol (L)-Tartrate, Dextromethorphan Hydrobromide Imp. B (EP) as (L)-Tartrate, Dextrorphan Tartrate, O-Desmethyldextromethorphan Tartrate, Dextromethorphan Hydrobromide Imp. B (EP), Dextromethorphan Imp. B (EP), Morphinan-3-ol, 17-methyl-, (9alpha,13alpha,14alpha)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), Morphinan-3-ol, 17-methyl-, (9alpha,13alpha,14alpha)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt) (9CI), Morphinan-3-ol, 17-methyl-, (9alpha,13alpha,14alpha)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), Dextrorphan bitartrate, NIH 4591, d-3-Hydroxy-N-methylmorphinan tartrate, Dextromethorphan Hydrobromide Impurity B as (L)-Tartrate, Dextromethorphan Impurity B as (L)-Tartrate |
| InChI formel | InChI=1S/C17H23NO.C4H6O6/c1-18-9-8-17-7-3-2-4-14(17)16(18)10-12-5-6-13(19)11-15(12)17;5-1(3(7)8)2(6)4(9)10/h5-6,11,14,16,19H,2-4,7-10H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t14-,16+,17+;1-,2-/m11/s1 |
| SMIL | CN1CC[C@@]23CCCC[C@@H]2[C@@H]1Cc4ccc(O)cc34.O[C@H]([C@@H](O)C(=O)O)C(=O)O |
| IUPAC navn | (9α,13α,14α)-3-Hydroxy-17-methylmorphinan (2R,3R)-2,3-dihydroxybutanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?