Få mere at vide
Dextrorphan-d3 b-D-O-Glucuronide, TRC
Artikelnummer. 30366043
Skift visning
Mængde:
2 mg
Artikelnummer. Mængde
30366043 2 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366043 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2995022MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dextrorphan-O-Glucuronide-D3
CAS 524713-58-4
Molekylær formel C23 2H3 H28 N O7
Mængde 2 mg
Synonym Dextrorphan-D3 B-D-O-Glucuronide
InChI formel InChI=1S/C23H31NO7/c1-24-9-8-23-7-3-2-4-14(23)16(24)10-12-5-6-13(11-15(12)23)30-22-19(27)17(25)18(26)20(31-22)21(28)29/h5-6,11,14,16-20,22,25-27H,2-4,7-10H2,1H3,(H,28,29)/t14-,16+,17-,18-,19+,20-,22+,23+/m0/s1/i1D3
SMIL [2H]C([2H])([2H])N1CC[C@]23CCCC[C@H]2[C@H]1Cc4ccc(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O)cc34
Molekylvægt (g/mol) 436.51
Formel vægt 436.2289