Få mere at vide
Dextropropoxyphene Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dextropropoxyphene Hydrochloride |
| CAS | 1639-60-7 |
| Analyt- eller komponentnavne | Dextropropoxyphene Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C22 H29 N O2 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Dextropropoxyphene Hydrochloride, (+)-4-(Dimethylamino)-3-methyl-1,2-diphenyl-2-butanol propionate (ester) hydrochloride, (2S,3R)-(+)-4-(Dimethylamino)-3-methyl-1,2-diphenyl-2-butanol propionate (ester) hydrochloride, (+)-Propoxyphene hydrochloride, Algaphan, Antalvic, Darvon, Darvon hydrochloride, Deprancol, Develin, Dextropropoxyphene hydrochloride, Dolene, Dolocap, Doloxen, Doloxene, Erantin, Femadol, Harmar, Propoxychel, Propoxyphen hydrochloride, Propoxyphene hydrochloride, Proxagesic, SK 65, d-4-Dimethylamino-3-methyl-1,2-diphenyl-2-butanol propionate hydrochloride, d-Propoxyphene hydrochloride, d-Propoxyphene monohydrochloride, (alphaS)-alpha-[(1R)-2-(Dimethylamino)-1-methylethyl]-alpha-phenylbenzeneethanol 1-propanoate hydrochloride (1:1), (alphaS)-alpha-[(1R)-2-(Dimethylamino)-1-methylethyl]-alpha-phenylbenzeneethanol propanoate (ester) hydrochloride, [S-(R*,S*)]-alpha-[2-(Dimethylamino)-1-methylethyl]-alpha-phenylbenzeneethanol propanoate (ester) hydrochloride, alpha-(+)-Propoxyphene hydrochloride, alpha-Propoxyphene hydrochloride, alpha-d-Propoxyphene hydrochloride |
| InChI formel | InChI=1S/C22H29NO2.ClH/c1-5-21(24)25-22(18(2)17-23(3)4,20-14-10-7-11-15-20)16-19-12-8-6-9-13-19;/h6-15,18H,5,16-17H2,1-4H3;1H/t18-,22+;/m1./s1 |
| Løsningstype | Neat |
| SMIL | Cl.CCC(=O)O[C@@](Cc1ccccc1)([C@H](C)CN(C)C)c2ccccc2 |
| Vis mere |