Få mere at vide
Dextromethorphan-d3 Hydrobromide, TRC
Artikelnummer. 30366275
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366275 50 mg
30363283 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30366275 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29945650MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1443149-12-9
Molekylær formel C18H23D3BrNO • H2O
Molekylvægt (g/mol) 355.331802
Synonym (9α,13α,14α)-3-Methoxy-d3-17-(methyl)morphinan Hydrobromide Hydrate, (+)-3-Methoxy-d3-N-(methyl-d3)morphinan Hydrobromide Hydrate, d-3-Methoxy-d3-N-(methyl-d3)morphinan Hydrobromide Hydrate
SMIL [H][C@@]1([C@@H](N(C)CC2)C3)[C@@]2(CCCC1)C4=C3C=CC(OC([2H])([2H])[2H])=C4.Br
Tekniske data
Kemisk navn eller materiale Dextromethorphan O-trideuteromethyl Hydrobromide Hydrate
Mængde 50 mg
InChI formel InChI=1S/C18H25NO.BrH/c1-19-10-9-18-8-4-3-5-15(18)17(19)11-13-6-7-14(20-2)12-16(13)18;/h6-7,12,15,17H,3-5,8-11H2,1-2H3;1H/t15-,17+,18+;/m1./s1/i2D3;