missing translation for 'onlineSavingsMsg'
Learn More

Dextrin, (Off-white to yellow powder/Commercial Grade Type II), MP Biomedicals

Product Code. 11445532 Shop All MP Biomedical Products
Click to view available options
Quantity:
3 kg
Unit Size:
3kg
This item is not returnable. View return policy
This item is not returnable. View return policy

80% water-soluble

TRUSTED_SUSTAINABILITY

Specifications

CAS 9004-53-9
Color White to Yellow
Molecular Formula C18H32O16
Quantity 3 kg
Synonym dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen
InChI Key FYGDTMLNYKFZSV-MRCIVHHJSA-N
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O
IUPAC Name (3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol) 504.438
PubChem CID 62698
Grade Commercial Type II
Physical Form Powder
Chemical Name or Material Dextrin, From Corn
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.