missing translation for 'onlineSavingsMsg'
Learn More

Dextran Sulfate, Sodium Salt, Ultrapure, Thermo Scientific™

Catalog Number 15855218
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
9011-18-1
(C6H7O5)m(C6H7O5)n
40,000
MFCD00081551
BLFLLBZGZJTVJG-UHFFFAOYSA-N
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
2337
CHEBI:116735
ethyl 4-aminobenzoate
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
This item is not returnable. View return policy
9011-18-1
(C6H7O5)m(C6H7O5)n
40,000
MFCD00081551
BLFLLBZGZJTVJG-UHFFFAOYSA-N
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
2337
CHEBI:116735
ethyl 4-aminobenzoate
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 10 g
Formula Weight 500,000 MW
Packaging Plastic bottle
Chemical Name or Material Dextran sulfate sodium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'