Learn More
Abcam Dextran Sulfate, Anhydroglucose Polymer, 500,000 g/mol, Solid, 10 g
Product Code. 32120450 Shop All Abcam Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity Unit Size
32120450 10 g 10g
1 options
Product Code. 32120450 Supplier Abcam Supplier No. ab14656910g
 Restricted Item
Forventet forsendelse: 24-11-2026
Anhydroglucose polymer used for probe hybridization and selective lipoprotein precipitation.
  • Molecular weight: 500,000 g/mol
  • Form: Solid
  • Composition: 100% Dextran sulfate
  • Applications: Selective precipitation of lipoproteins, probe hybridization of membrane-immobilized DNA, release of DNA from DNA-histone complexes, and inhibition of RNA binding to ribosomes
  • Key Benefit: Accelerates the rate of probe hybridization by 10 fold when used as an additive
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Buffer 100% Dextran sulfate
Content And Storage Store at 4°C.
Form Solid
Shipping Condition Ambient
Target Molecule Lipoproteins, DNA, RNA
Formulation 100% Dextran sulfate
Quantity 10 g
For Use With (Application) Selective precipitation of lipoproteins, probe hybridization of membrane-immobilized DNA, release of DNA from DNA-histone complexes, inhibition of RNA binding to ribosomes