Få mere at vide
Dexpropranolol, LoGiCal
Artikelnummer. 30459419
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459419 10 mg
1 options
Artikelnummer. 30459419 Leverandør LoGiCal Leverandørnr. LGCFOR3350.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dexpropranolol
CAS 5051-22-9
Analyt- eller komponentnavne Dexpropranolol
Forsendelsestilstand Room Temperature
Molekylær formel C16 H21 N O2
Certificeringer/compliance ISO 9001
Synonym 2-Propanol, 1-[(1-methylethyl)amino]-3-(1-naphthalenyloxy)-, (2R)-, 2-Propanol, 1-(isopropylamino)-3-(1-naphthyloxy)-, (+)- (8CI), 2-Propanol, 1-[(1-methylethyl)amino]-3-(1-naphthalenyloxy)-, (R)-, (+)-(R)-Propranolol, (+)-Propranolol, (2R)-Propranolol, (R)-(+)-Propranolol, (R)-Propranolol, Dexpropranolol, Dextropropranolol, d-Propranolol
InChI formel InChI=1S/C16H21NO2/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16/h3-9,12,14,17-18H,10-11H2,1-2H3/t14-/m1/s1
Løsningstype Neat
SMIL CC(C)NC[C@@H](O)COc1cccc2ccccc12
IUPAC navn (2R)-1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol
Industry Type Forensics
Molekylvægt (g/mol) 259.34
Mængde 10 mg
Formel vægt 259.1572 g/mol