missing translation for 'onlineSavingsMsg'
Få mere at vide

Dexmedetomidine hydrochloride, MedChemExpress

Artikelnummer. 16449797
Encompass_Preferred
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
Denne vare kan ikke returneres. Se returpolicy

Artikelnummer. 16449797

Brand: Medchem Express HY17034A

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Dexmedetomidine hydrochloride ((+)-Medetomidine hydrochloride) is a potent, selective and orally active agonist of α2-adrenoceptor, with a Ki of 1.08 nM. Dexmedetomidine hydrochloride shows 1620-fold selectivity against α1-adrenoceptor. Dexmedetomidine hydrochloride exhibits anxiolysis, sedation, and modest analgesia effects.

Dexmedetomidine hydrochloride ((+)-Medetomidine hydrochloride) is a potent, selective and orally active agonist of α2-adrenoceptor, with a Ki of 1.08 nM. Dexmedetomidine hydrochloride shows 1620-fold selectivity against α1-adrenoceptor. Dexmedetomidine hydrochloride exhibits anxiolysis, sedation, and modest analgesia effects.

Tekniske data

Kemisk navn eller materiale Dexmedetomidine hydrochloride
Fysisk form Solid
CAS 145108-58-3
Farve White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C13H17ClN2
Til brug med (applikation) Cancer-programmed cell death
Synonym (+)-Medetomidine hydrochloride(S)-Medetomidine hydrochloride
Opløselighedsinformation DMSO : 250 mg/mL (1056.01 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (211.20 mM)
SMIL C[C@H](C1=CN=CN1)C2=CC=CC(C)=C2C.Cl
Molekylvægt (g/mol) 236.74
Formel vægt 236.74
Procent renhed 99.39%
Grad Research
Vis mere Vis mindre

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.