missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dexketoprofen Ethyl Ester, Mikromol™
Description
Dexketoprofen Ethyl Ester
Specifications
Specifications
| Kemisk navn eller materiale | Dexketoprofen Ethyl Ester |
| CAS | 136656-97-8 |
| Analyt- eller komponentnavne | Dexketoprofen Ethyl Ester |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C18 H18 O3 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Dexketoprofen Ethyl Ester, (alphaS)-3-Benzoyl-alpha-methylbenzeneacetic acid ethyl ester, Benzeneacetic acid, 3-benzoyl-alpha-methyl-, ethyl ester, (S)-, (S)-Ketoprofen ethyl ester |
| InChI formel | InChI=1S/C18H18O3/c1-3-21-18(20)13(2)15-10-7-11-16(12-15)17(19)14-8-5-4-6-9-14/h4-13H,3H2,1-2H3/t13-/m0/s1 |
| SMIL | CCOC(=O)[C@@H](C)c1cccc(c1)C(=O)c2ccccc2 |
| IUPAC navn | ethyl (2S)-2-(3-benzoylphenyl)propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?