missing translation for 'onlineSavingsMsg'
Få mere at vide

Dexibuprofen Isopropyl Ester, Mikromol™

Product Code. 17983356
Change view
Click to view available options
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
17983356 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17983356 Supplier Mikromol Supplier No. MM3534.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dexibuprofen Isopropyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Dexibuprofen Isopropyl Ester
CAS 1114975-39-1
Analyt- eller komponentnavne Dexibuprofen Isopropyl Ester
Forsendelsestilstand Room Temperature
Molekylær formel C16 H24 O2
Certificeringer/compliance ISO 9001
Synonym Dexibuprofen Isopropyl Ester, (alphaS)-alpha-Methyl-4-(2-methylpropyl)benzeneacetic acid 1-methylethyl ester
InChI formel InChI=1S/C16H24O2/c1-11(2)10-14-6-8-15(9-7-14)13(5)16(17)18-12(3)4/h6-9,11-13H,10H2,1-5H3/t13-/m0/s1
SMIL CC(C)Cc1ccc(cc1)[C@H](C)C(=O)OC(C)C
IUPAC navn propan-2-yl (2S)-2-[4-(2-methylpropyl)phenyl]propanoate
Molekylvægt (g/mol) 248.36
Mængde 100 mg
Formel vægt 248.1776 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.