Få mere at vide
Dexetimide, MedChemExpress
Artikelnummer. 16484038
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16484038 10 mg
16430397 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16484038 Leverandør Medchem Express Leverandørnr. HY105545
MedChemExpress Dexetimide ((+)-Benzetimide) is a high-affinity muscarinic receptor antagonist and a potent and persistent anticholinergic agent used to treat neuroleptic-induced parkinsonism.
Dexetimide ((+)-Benzetimide) is a high-affinity muscarinic receptor antagonist and a potent and persistent anticholinergic agent used to treat neuroleptic-induced parkinsonism.
Tekniske data
Kemisk navn eller materiale Dexetimide
CAS 21888-98-2
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C23H26N2O2
Mængde 10 mg
Synonym (+)-Benzetimide (S)-(+)-Dexetimide Dexbenzetimide
Opløselighedsinformation DMSO : ≥ 100 mg/mL (275.89 mM)
SMIL O=C(N1)[C@@](C2CCN(CC3=CC=CC=C3)CC2)(C4=CC=CC=C4)CCC1=O
Molekylvægt (g/mol) 362.46
Formel vægt 362.46
Procent renhed 99.2%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation