Få mere at vide
Dexchlorpheniramine Maleate, Mikromol™
Beskrivelse
Dexchlorpheniramine Maleate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Dexchlorpheniramine Maleate |
| CAS | 2438-32-6 |
| Analyt- eller komponentnavne | Dexchlorpheniramine Maleate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H19 Cl N2 . C4 H4 O4 |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Brompheniramine Maleate Imp. B (EP), Brompheniramine Imp. B (EP), Dexchlorpheniramine Maleate, Brompheniramine Maleate Imp. B (EP) as Maleate, Brompheniramine Maleate Impurity B as Maleate, Brompheniramine Impurity B as Maleate |
| SMIL | CN(C)CC[C@@H](c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| Molekylvægt (g/mol) | 390.86 |
| Mængde | 100 mg |
| Vis mere |