Få mere at vide
Dexamethasone-d5, TRC
Artikelnummer. 30361331
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30361331 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361331 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29880210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dexamethasone-D5
CAS 358731-91-6
Renhedsgrad noter HPLC
Molekylær formel C22 2H5 H24 F O5
Mængde 10 mg
Synonym Dexamethasone D5 (4,6,6,21,21 D5), Pregna-1,4-diene-3,20-dione-4,6,6,21,21-d5, 9-fluoro-11,17,21-trihydroxy-16-methyl-, (11β,16α)- (9CI), Dexamethasone-d5, Dexamethasone-4,6,6,21,21-D5, (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione-4,6,6,21,21-d5, 9-Fluoro-11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione-4,6,6,21,21-d5
InChI formel InChI=1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1/i4D2,9D,11D2
SMIL [2H]C1=C2C([2H])([2H])C[C@H]3[C@@H]4C[C@@H](C)[C@](O)(C(=O)C([2H])([2H])O)[C@@]4(C)C[C@H](O)[C@]3(F)[C@@]2(C)C=CC1=O
IUPAC navn (8S,9R,10S,11S,13S,14S,16R,17R)-4,6,6-trideuterio-17-(2,2-dideuterio-2-hydroxyacetyl)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-8,11,12,14,15,16-hexahydro-7H-cyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 397.49
Formel vægt 397.2313
Procent renhed >95