Få mere at vide
Dexamethasone-biotin, TRC
Artikelnummer. 30361837
Skift visning
Mængde:
2.5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361837 100 mg
30359514 10 mg
30361753 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30361837 Leverandør Toronto Research Chemicals Leverandørnr. TRCD298765100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85679-56-7
Molekylær formel C36 H53 F N4 O6 S
Molekylvægt (g/mol) 688.89
Synonym N-[5-[[[(11β,16α,17α)-9-Fluoro-11,17-dihydroxy-16-methyl-3-oxoandrosta-1,4-dien-17-yl]carbonyl]amino]pentyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide
IUPAC navn (8S,9R,10S,11S,13S,14S,16R,17R)-N-[5-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]pentyl]-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxamide
SMIL C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)NCCCCCNC(=O)CCCC[C@@H]5SC[C@@H]6NC(=O)N[C@H]56
Tekniske data
Kemisk navn eller materiale Dexamethasone-biotin
Mængde 100 mg
InChI formel InChI=1S/C36H53FN4O6S/c1-21-17-25-24-12-11-22-18-23(42)13-14-33(22,2)35(24,37)28(43)19-34(25,3)36(21,47)31(45)39-16-8-4-7-15-38-29(44)10-6-5-9-27-30-26(20-48-27)40-32(46)41-30/h13-14,18,21,24-28,30,43,47H,4-12,15-17,19-20H2,1-3H3,(H,38,44)(H,39,45)(H2,40,41,46)/t21-,24+,25+,26+,27+,28+,30+,33+,34+,35+,36+/m1/s1
Formel vægt 688.367