Learn More
Dexamethasone 9,11-epoxide, MedChemExpress Product Code.: 16422376
Dexamethasone 9,11-epoxide, MedChemExpress
Product Code. 16422376
500 mg, 500mg
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
Product Code. 16422376
Brand: Medchem Express HYN0348
MedChemExpress Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.
Dexamethasone 9,11-epoxide, a compound extracted from patent CN 106520896 A and RU 2532902 C1, is an intermediate in the preparation of dexamethasone.
Specifications
Chemical Name or Material Dexamethasone 9,11-epoxide
CAS 24916-90-3
Color White
Purity Grade Notes Research
Molecular Formula C22H28O5
Quantity 500 mg
Solubility Information DMSO : 100 mg/mL (268.49 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@@]12[C@](C(CO)=O)(O)[C@H](C)C[C@@]1([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@]3(O5)[C@]5([H])C2)=O
Molecular Weight (g/mol) 372.45
Formula Weight 372.45
Percent Purity 99.3%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Dexamethasone 9,11-epoxide, MedChemExpress