Få mere at vide
Dexamethasone 21-Propionate, TRC
Artikelnummer. 30362642
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30362642 25 mg -
30353783 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30362642 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29882525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3793-10-0
Molekylær formel C25 H33 F O6
Molekylvægt (g/mol) 448.52
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11,17-dihydroxy-16-methyl-21-(1-oxopropoxy)-, (11β,16α)-, Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-, 21-propionate (7CI,8CI), (11β,16α)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Dexamethasone 21-Propionate
IUPAC navn [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
SMIL CCC(=O)OCC(=O)[C@@]1(O)[C@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
Tekniske data
Kemisk navn eller materiale Dexamethasone 21-Propionate
Mængde 25 mg
InChI formel InChI=1S/C25H33FO6/c1-5-21(30)32-13-20(29)25(31)14(2)10-18-17-7-6-15-11-16(27)8-9-22(15,3)24(17,26)19(28)12-23(18,25)4/h8-9,11,14,17-19,28,31H,5-7,10,12-13H2,1-4H3/t14-,17+,18+,19+,22+,23+,24+,25+/m1/s1
Formel vægt 448.2261