missing translation for 'onlineSavingsMsg'
Få mere at vide

Dexamethasone 21-Propionate, TRC

Artikelnummer. 30353783
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30353783 2.5 mg 2.5mg
30362642 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30353783 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2988252.5MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 3793-10-0
Molekylær formel C25 H33 F O6
Molekylvægt (g/mol) 448.52
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11,17-dihydroxy-16-methyl-21-(1-oxopropoxy)-, (11β,16α)-, Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-, 21-propionate (7CI,8CI), (11β,16α)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Dexamethasone 21-Propionate
IUPAC navn [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
SMIL CCC(=O)OCC(=O)[C@@]1(O)[C@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C

Specifications

Kemisk navn eller materiale Dexamethasone 21-Propionate
Mængde 2.5 mg
InChI formel InChI=1S/C25H33FO6/c1-5-21(30)32-13-20(29)25(31)14(2)10-18-17-7-6-15-11-16(27)8-9-22(15,3)24(17,26)19(28)12-23(18,25)4/h8-9,11,14,17-19,28,31H,5-7,10,12-13H2,1-4H3/t14-,17+,18+,19+,22+,23+,24+,25+/m1/s1
Formel vægt 448.2261
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.