Learn More
Devarda's Alloy, For Analysis, EMSURE™ MilliporeSigma™
Product Code. 15854550
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
Unit Size:
250g
Product Code. Quantity Packaging Unit Size
15854550 250 g Plastic Bottle 250g
1 options
Product Code. 15854550 Supplier Supelco Supplier No. 1053410250
 Restricted Item
Forventet forsendelse: 19-11-2026
Recommended Storage
+5°C to +30°C.
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
CAS 8049-11-4
Melting Point 490°C to 560°C
Boiling Point 906°C
Density 5.79 g/3 (20 °C)
Quantity 250 g
Packaging Plastic Bottle
Form Solid
Molecular Formula Al9Cu10Zn
Synonym devarda's alloy, devarda's alloy, granular, al.cu.zn, devarda's alloy, p.a., alumane copper zinc, devarda's alloy, puriss. p.a., devarda's alloy, saj special grade, devarda's alloy, powder,-100 mesh, devarda's alloy, filings, +100 mesh, devarda inverted exclamation mark s alloy
InChI Key MPKKOQSNGWAMJD-UHFFFAOYSA-N
SMILES [Al].[Cu].[Zn]
IUPAC Name aluminum;copper;zinc
Molecular Weight (g/mol) 943.67
PubChem CID 16211762