Få mere at vide
Deterenol-d7 Hydrochloride, TRC
Artikelnummer. 30354003
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30354003 25 mg
1 options
Artikelnummer. 30354003 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29795225MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Deterenol-d7 Hydrochloride
CAS 1246817-36-6
Molekylær formel C11H10D7NO2 • HCl
Mængde 25 mg
Synonym 4-Hydroxy-α-[[(1-methylethyl-d7)amino]methyl]benzenemethanol Hydrochloride, (+/-)-p-Hydroxy-α-[(isopropyl-d7-amino)methyl]benzyl Alcohol Hydrochloride, (+/-)-1-(4-Hydroxyphenyl)-2-(isopropyl-d7)aminoethanol Hydrochloride, Detenerol-d7 Hydrochloride, dl-1-(4-Hydroxyphenyl)-2-(isopropyl-d7-amino)ethanol Hydrochloride
InChI formel InChI=1S/C11H17NO2.ClH/c1-8(2)12-7-11(14)9-3-5-10(13)6-4-9;/h3-6,8,11-14H,7H2,1-2H3;1H/i1D3,2D3,8D;
SMIL OC1=CC=C(C(O)CNC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1.Cl
Molekylvægt (g/mol) 202.303646