Få mere at vide
Cole-Parmer™ Botulinum Toxin Detection Kit
Artikelnummer. 13234493 Shop alle Cole-Parmer produkter
Skift visning
For use with: Toxin Detection.
Artikelnummer.
13234493 For use with: Toxin Detection.
1 options
Artikelnummer. 13234493 Leverandør Cole-Parmer™ Leverandørnr. 3610814
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Accurately detect small, yet high threat amounts of biowarfare agents-without extensive sample preparation or additional expensive equipment.
  • Results in 3- 30 minutes
  • Easy to read – two lines for positive; one line for negative
  • High Sensitivity and specificity
  • Biohazard disposal bags and chain of custody labels are included for proper control and disposal or transport of the sample and test
Tekniske data
Oversigt Botulinum Toxin Detection Kit, handheld assay, 10/bx
Test Type Botulinum Toxin
Mængde 10/Box