Få mere at vide
Desulfovardenafil (2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one), Mikromol™
Artikelnummer. 30049509
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049509 100 mg
30049498 25 mg
30458484 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30049509 Leverandør Mikromol Leverandørnr. MM1569.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Desulfovardenafil (2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one)
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desulfovardenafil (2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one)
CAS 224789-21-3
Analyt- eller komponentnavne Desulfovardenafil (2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one)
Forsendelsestilstand Room Temperature
Molekylær formel C17 H20 N4 O2
Certificeringer/compliance ISO 17025
Synonym Imidazo[5,1-f][1,2,4]triazin-4(1H)-one, 2-(2-ethoxyphenyl)-5-methyl-7-propyl-, 2-(2-Ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one
InChI formel InChI=1S/C17H20N4O2/c1-4-8-14-18-11(3)15-17(22)19-16(20-21(14)15)12-9-6-7-10-13(12)23-5-2/h6-7,9-10H,4-5,8H2,1-3H3,(H,19,20,22)
SMIL CCCc1nc(C)c2C(=O)NC(=Nn12)c3ccccc3OCC
IUPAC navn 2-(2-ethoxyphenyl)-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one
Molekylvægt (g/mol) 312.37
Mængde 100 mg
Formel vægt 312.1586 g/mol