missing translation for 'onlineSavingsMsg'
Få mere at vide

Desulfonamide MLN 4924, TRC

Artikelnummer. 30354878
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30354878 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30354878 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29792010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale (1S,2S,4R)-4-(4-(((1S)-2,3-Dihydro-1H-inden-1-yl)amino)-7H-pyrrolo(2,3-d)pyrimidin-7-yl)-2-hydroxy-cyclopentanemethanol
CAS 905580-90-7
Molekylær formel C21H24N4O2
Mængde 10 mg
Synonym Desulfonamide MLN 4924, (1S,2S,4R)-4-[4-[[(1S)-2,3-Dihydro-1H-inden-1-yl]amino]-7H-pyrrolo[2,3-d]pyrimidin-7-yl]-2-hydroxy-cyclopentanemethanol
InChI formel InChI=1S/C21H24N4O2/c26-11-14-9-15(10-19(14)27)25-8-7-17-20(22-12-23-21(17)25)24-18-6-5-13-3-1-2-4-16(13)18/h1-4,7-8,12,14-15,18-19,26-27H,5-6,9-11H2,(H,22,23,24)/t14-,15+,18-,19-/m0/s1
SMIL OC[C@@H]1C[C@H](C[C@@H]1O)n2ccc3c(N[C@H]4CCc5ccccc45)ncnc23
IUPAC navn (1S,2S,4R)-4-[4-[[(1S)-2,3-dihydro-1H-inden-1-yl]amino]pyrrolo[2,3-d]pyrimidin-7-yl]-2-(hydroxymethyl)cyclopentan-1-ol
Molekylvægt (g/mol) 364.44
Formel vægt 364.1899
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.