Få mere at vide
Desphenyl Chloridazon-15N2, TRC
Artikelnummer. 17903665
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17903665 10 mg
17943665 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17903665 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29700210MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1189649-21-5
Molekylær formel C4 H4 Cl 15N2 N O
Molekylvægt (g/mol) 147.53
Synonym Chloridazon Impurity 1-15N2
IUPAC navn 4-amino-5-chloro-(1,2-^{15}N_{2})1H-pyridazin-6-one
SMIL NC1=C(Cl)C(=O)[15NH][15N]=C1
Tekniske data
Kemisk navn eller materiale Chloridazon-desphenyl 15N2
Mængde 10 mg
InChI formel InChI=1S/C4H4ClN3O/c5-3-2(6)1-7-8-4(3)9/h1H,(H3,6,8,9)/i7+1,8+1
Formel vægt 146.9984