Få mere at vide
Desoxycarbadox-D3, TRC
Artikelnummer. 30350249
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30350249 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30350249 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2969971MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desoxycarbadox methyl-d3
CAS 1448350-02-4
Renhedsgrad noter HPLC
Molekylær formel C11 2H3 H7 N4 O2
Mængde 1 mg
Synonym Trideuteriomethyl N-(quinoxalin-2-ylmethylideneamino)carbamate
InChI formel InChI=1S/C11H10N4O2/c1-17-11(16)15-13-7-8-6-12-9-4-2-3-5-10(9)14-8/h2-7H,1H3,(H,15,16)/b13-7+/i1D3
SMIL [2H]C([2H])([2H])OC(=O)N\N=C\c1cnc2ccccc2n1
IUPAC navn trideuteriomethyl N-[(E)-quinoxalin-2-ylmethylideneamino]carbamate
Molekylvægt (g/mol) 233.24
Formel vægt 233.0992
Procent renhed >95