Få mere at vide
Desoxo-palbociclib, TRC
Artikelnummer. 30356862
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356862 10 mg
30351286 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30356862 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29266010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2204863-06-7
Molekylær formel C24H29N7O
Molekylvægt (g/mol) 431.53
Synonym Palbociclib Impurity-2, Palbociclib Impurity D
SMIL CC1=C(C=C)C(=O)N(C2CCCC2)c3nc(Nc4ccc(cn4)N5CCNCC5)ncc13
Tekniske data
Kemisk navn eller materiale Desoxo-palbociclib
Mængde 10 mg
InChI formel InChI=1S/C24H29N7O/c1-3-19-16(2)20-15-27-24(29-22(20)31(23(19)32)17-6-4-5-7-17)28-21-9-8-18(14-26-21)30-12-10-25-11-13-30/h3,8-9,14-15,17,25H,1,4-7,10-13H2,2H3,(H,26,27,28,29)