Få mere at vide
Desogestrel, MedChemExpress
Artikelnummer. 16485228
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16485228 10 mg 10mg
16421447 50 mg 50mg
16445906 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16485228 Leverandør Medchem Express Leverandørnr. HY12516
MedChemExpress Desogestrel(Org-2969) is a third-generation 19-nortestosterone derivative progestogen; is contained in many oral contraceptive preparations, both combined (COCs) to ethinyl-estradiol (EE) or alone in a progestin-only pill (POP).
Desogestrel(Org-2969) is a third-generation 19-nortestosterone derivative progestogen
Tekniske data
Kemisk navn eller materiale Desogestrel
Fysisk form Solid
CAS 54024-22-5
Farve White
Mængde 10 mg
Renhedsgrad noter Research
Molekylær formel C22H30O
Synonym Org-2969
Opløselighedsinformation DMSO : 16.67 mg/mL (53.69 mM; Need ultrasonic)
SMIL [H][C@@]12C(CC[C@]([C@@](CC[C@@]3(O)C#C)([H])[C@]3(CC)C4)([H])[C@]2([H])C4=C)=CCCC1
Molekylvægt (g/mol) 310.47
Formel vægt 310.47
Procent renhed 99.7%