Få mere at vide
Desmosterol-d6, TRC
Artikelnummer. 30358865
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30358865 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30358865 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29686210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desmosterol 26,26,26,27,27,27-d6
CAS 1246298-67-8
Molekylær formel C27 2H6 H38 O
Mængde 10 mg
Synonym Cholesta-5,24-dien-26,26,26,27,27,27-d6-3-ol, (3β)-, (3β)-Cholesta-5,24-dien-26,26,26,27,27,27-d6-3-ol, Desmosterol-d6, Desmosterol-26,27-d6, Desmosterol-D6 (26,26,26,27,27,27-D6), 24,25-Dehydrocholesterol-26,27-d6, 24-Dehydrocholesterol-d6
InChI formel InChI=1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h7,9,19,21-25,28H,6,8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i1D3,2D3
SMIL [2H]C([2H])([2H])C(=CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C([2H])([2H])[2H]
IUPAC navn (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-7,7,7-trideuterio-6-(trideuteriomethyl)hept-5-en-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
Molekylvægt (g/mol) 390.67
Formel vægt 390.3769